Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

MES monohydrate, 99+%, for biochemistry

Catalog number: 327760025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

MES monohydrate, 99+%, for biochemistry

Catalog number: 327760025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 327760025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
145224-94-8
IUPAC Name
4-(2-sulfonatoethyl)morpholin-4-ium
Molecular Formula
C6H13NO4S
InChI Key
SXGZJKUKBWWHRA-UHFFFAOYSA-N
SMILES
[O-]S(=O)(=O)CC[NH+]1CCOCC1
Appearance (Color)
White
Titration with NaOH
99.0 to 101.0 %
Water
8.0 to 9.0 % (K.F.)
pH
2.5 to 4.0 (1 % in water at 25°C)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text