Tricine, 99+%, for biochemistry
This product ships via ground from US northeast.
Tricine, 99+%, for biochemistry
Thermo Scientific Chemicals

Tricine, 99+%, for biochemistry

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
25 g
100 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 327830050
Price (USD)/ Each
1,849.65
Online exclusive
2176.00 
Save 326.35 (15%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
1,849.65
Online exclusive
2176.00 
Save 326.35 (15%)
Add to cart
Tricine, 99+%, for biochemistry
Catalog number327830050
Price (USD)/ Each
1,849.65
Online exclusive
2176.00 
Save 326.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}acetic acid
Molecular FormulaC6H13NO5
InChI KeySEQKRHFRPICQDD-UHFFFAOYSA-N
SMILESOCC(CO)(CO)NCC(O)=O
Molecular Weight (g/mol)179.17
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=99.0 %
Water=<1 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only