n-Butyl acetate, 99.5+%, for electronic use (MOS), residue free, Thermo Scientific Chemicals
n-Butyl acetate, 99.5+%, for electronic use (MOS), residue free, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Butyl acetate, 99.5+%, for electronic use (MOS), residue free, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 L
Catalog number 327850010
Price (USD)/ Each
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
IUPAC Namebutyl acetate
Molecular FormulaC6H12O2
InChI KeyDKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILESCCCCOC(C)=O
Molecular Weight (g/mol)116.16
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<0.02 ppm
Appearance (Form)Clear liquid
Water=<0.1 % (Coulometric)
Heavy metals (as Pb)=<50 ppm
Arsenic (As)=<0.01 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For electronic use
RUO – Research Use Only