Thermo Scientific Chemicals

Methanol, 99.8%, for electronic use, residue free, Thermo Scientific Chemicals

Catalog number: 327900010
1 L, Glass Bottle, Each
Thermo Scientific Chemicals

Methanol, 99.8%, for electronic use, residue free, Thermo Scientific Chemicals

Catalog number: 327900010
1 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 327900010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
67-56-1
IUPAC Name
methanol
Molecular Formula
CH4O
InChI Key
OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES
CO
Appearance (Form)
Clear liquid
Chloride (Cl)
=<2 ppm
Water
=<0.1 % (Coulometric)
Arsenic (As)
=<50 ppb
Copper (Cu)
=<50 ppb

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For electronic use
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text