AEBSF hydrochloride, 98%
AEBSF hydrochloride, 98%
Thermo Scientific Chemicals

AEBSF hydrochloride, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
1 g
Packaging:
Glass bottle
Catalog number 328110500
Price (USD)/ Each
70.65
Online exclusive
83.50 
Save 12.85 (15%)
-
Add to cart
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
70.65
Online exclusive
83.50 
Save 12.85 (15%)
Add to cart
AEBSF hydrochloride, 98%
Catalog number328110500
Price (USD)/ Each
70.65
Online exclusive
83.50 
Save 12.85 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namehydrogen 4-(2-aminoethyl)benzene-1-sulfonyl fluoride chloride
Molecular FormulaC8H11ClFNO2S
InChI KeyWRDABNWSWOHGMS-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCCC1=CC=C(C=C1)S(F)(=O)=O
Molecular Weight (g/mol)239.69
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only