Chemical Identifiers
CAS97161-97-2
IUPAC Namemethyl 16-hydroxy-15-methyl-3-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8(13),9,11,20,22,24-nonaene-16-carboxylate
Molecular FormulaC27H21N3O5
InChI KeyKOZFSFOOLUUIGY-UHFFFAOYNA-N
SMILESCOC(=O)C1(O)CC2OC1(C)N1C3=C(C=CC=C3)C3=C1C1=C(C4=CC=CC=C4N21)C1=C3CNC1=O
View more