Thermo Scientific Chemicals

Okadaic acid, 97%, Thermo Scientific Chemicals

Catalog number: 328370001
0.1 mg, Each
Thermo Scientific Chemicals

Okadaic acid, 97%, Thermo Scientific Chemicals

Catalog number: 328370001
0.1 mg, Each
Quantity
Catalog number: 328370001
Price (USD)
Have Questions?

Chemical Identifiers

CAS
78111-17-8
IUPAC Name
(2R)-3-[(2S,5R,6R,8S)-8-[(2R,3E)-4-[(2R,4'aR,5R,6'S,8'R)-8'-hydroxy-6'-[(1S,3S)-1-hydroxy-3-[(3R,6S)-3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl]butyl]-7'-methylidene-hexahydro-3'H-spiro[oxolane-2,2'-pyrano[3,2-b]pyran]-5-yl]but-3-en-2-yl]-5-hydroxy-10-methyl-1,7-dioxaspiro[5.5]undec-10-en-2-yl]-2-hydroxy-2-methylpropanoic acid
Molecular Formula
C44H68O13
InChI Key
QNDVLZJODHBUFM-AAWJMCDUSA-N
SMILES
C[C@@H](C[C@H](O)[C@H]1O[C@@H]2CC[C@@]3(CC[C@@H](O3)\C=C\[C@@H](C)[C@@H]3CC(C)=C[C@@]4(O[C@H](C[C@@](C)(O)C(O)=O)CC[C@H]4O)O3)OC2[C@H](O)C1=C)C1O[C@@]2(CCCCO2)CC[C@H]1C
Appearance (Form)
Solid
Appearance (Color)
White to off-white
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text