Piceatannol, 96%, Thermo Scientific™
Piceatannol, 96%, Thermo Scientific™
Thermo Scientific™

Piceatannol, 96%, Thermo Scientific™

Have Questions?
Catalog number 328450010
Price (USD)
-
Request bulk or custom format
Chemical Identifiers
CAS10083-24-6
IUPAC Name5-[(1E)-2-(3,4-dihydroxyphenyl)ethenyl]benzene-1,3-diol
Molecular FormulaC14H12O4
InChI KeyCDRPUGZCRXZLFL-OWOJBTEDSA-N
SMILESOC1=CC(C=CC2=CC=C(O)C(O)=C2)=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown-pink
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
Ask our AI about this Product
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection