Thermo Scientific™

Piceatannol

Catalog number: 328452500
250 mg, Each
Thermo Scientific™

Piceatannol

Catalog number: 328452500
250 mg, Each
Quantity
Catalog number: 328452500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10083-24-6
IUPAC Name
5-[(1E)-2-(3,4-dihydroxyphenyl)ethenyl]benzene-1,3-diol
Molecular Formula
C14H12O4
InChI Key
CDRPUGZCRXZLFL-OWOJBTEDSA-N
SMILES
OC1=CC(C=CC2=CC=C(O)C(O)=C2)=CC(O)=C1
Appearance (Color)
Yellow to brown-pink
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
HPLC
>=97.5 %

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text