Thermo Scientific Chemicals

Thapsigargin, 97%, Thermo Scientific Chemicals

Catalog number: 328570010
1 mg, Each
Thermo Scientific Chemicals

Thapsigargin, 97%, Thermo Scientific Chemicals

Catalog number: 328570010
1 mg, Each
Quantity
Catalog number: 328570010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67526-95-8
IUPAC Name
(3S,3aR,4S,6S,6aR,7S,8S,9bS)-6-(acetyloxy)-4-(butanoyloxy)-3,3a-dihydroxy-3,6,9-trimethyl-8-{[(2Z)-2-methylbut-2-enoyl]oxy}-2-oxo-2H,3H,3aH,4H,5H,6H,6aH,7H,8H,9bH-azuleno[4,5-b]furan-7-yl octanoate
Molecular Formula
C34H50O12
InChI Key
IXFPJGBNCFXKPI-FSIHEZPISA-N
SMILES
CCCCCCCC(=O)O[C@@H]1[C@@H](OC(=O)C(\C)=C/C)C(C)=C2[C@@H]3OC(=O)[C@@](C)(O)[C@@]3(O)[C@H](C[C@](C)(OC(C)=O)[C@@H]12)OC(=O)CCC
Appearance (Color)
Colorless to white to light yellow
Appearance (Form)
Powder or film
TLC
>=96.0 %
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text