Wortmannin, 98%
Wortmannin, 98%
Wortmannin, 98%
Thermo Scientific Chemicals

Wortmannin, 98%

CAS: 19545-26-7 | C23H24O8 | 428.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
5 mg
Catalog number 328590010
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
In stock
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS19545-26-7
IUPAC Name(1R,3R,5S,9R,18S)-18-(methoxymethyl)-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo[10.6.1.0²,¹⁰.0⁵,⁹.0¹⁵,¹⁹]nonadeca-2(10),12(19),14-trien-3-yl acetate
Molecular FormulaC23H24O8
InChI KeyQDLHCMPXEPAAMD-QAIWCSMKSA-N
SMILESCOC[C@H]1OC(=O)C2=COC3=C2[C@@]1(C)C1=C([C@@H]2CCC(=O)[C@@]2(C)C[C@H]1OC(C)=O)C3=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
HPLC>=97.5 %
Appearance (Form)Powder or solid or film
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471426Certificate of AnalysisMay 17, 2025328590010, 328590050
A0467289Certificate of AnalysisNov 07, 2024328590010, 328590050
A0464664Certificate of AnalysisJul 22, 2024328590010, 328590050
A0358804Certificate of AnalysisApr 12, 2024328590010, 328590050
A0461067Certificate of AnalysisMar 15, 2024328590010, 328590050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection