Thermo Scientific Chemicals

Wortmannin, 98%, Thermo Scientific Chemicals

Catalog number: 328590050
5 mg, Each
Thermo Scientific Chemicals

Wortmannin, 98%, Thermo Scientific Chemicals

Catalog number: 328590050
5 mg, Each
Quantity
Catalog number: 328590050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
19545-26-7
IUPAC Name
(1R,3R,5S,9R,18S)-18-(methoxymethyl)-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo[10.6.1.0²,¹⁰.0⁵,⁹.0¹⁵,¹⁹]nonadeca-2(10),12(19),14-trien-3-yl acetate
Molecular Formula
C23H24O8
InChI Key
QDLHCMPXEPAAMD-QAIWCSMKSA-N
SMILES
COC[C@H]1OC(=O)C2=COC3=C2[C@@]1(C)C1=C([C@@H]2CCC(=O)[C@@]2(C)C[C@H]1OC(C)=O)C3=O
Appearance (Color)
White to light yellow
HPLC
>=97.5 %
Appearance (Form)
Powder or solid or film

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text