DETA NONOate, 95%, Thermo Scientific Chemicals
DETA NONOate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DETA NONOate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Packaging:
Glass bottle
Catalog number 328650250
Price (USD)/ Each
89.65
Online exclusive
98.90 
Save 9.25 (9%)
-
Add to cart
Quantity:
25 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
89.65
Online exclusive
98.90 
Save 9.25 (9%)
Add to cart
DETA NONOate, 95%, Thermo Scientific Chemicals
Catalog number328650250
Price (USD)/ Each
89.65
Online exclusive
98.90 
Save 9.25 (9%)
-
Add to cart
Chemical Identifiers
CAS146724-94-9
IUPAC NameN-[bis(2-azaniumylethyl)amino]-N-nitrosohydroxylamine
Molecular FormulaC4H15N5O2
InChI KeyHMRRJTFDJAVRMR-UHFFFAOYSA-P
SMILES[NH3+]CCN(CC[NH3+])N(O)N=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline solid
Infrared spectrumConforms
NMR>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only