Thermo Scientific Chemicals

DETA NONOate, 95%, Thermo Scientific Chemicals

Catalog number: 328651000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

DETA NONOate, 95%, Thermo Scientific Chemicals

Catalog number: 328651000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 328651000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
146724-94-9
IUPAC Name
N-[bis(2-azaniumylethyl)amino]-N-nitrosohydroxylamine
Molecular Formula
C4H15N5O2
InChI Key
HMRRJTFDJAVRMR-UHFFFAOYSA-P
SMILES
[NH3+]CCN(CC[NH3+])N(O)N=O
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline solid
Infrared spectrum
Conforms
NMR
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text