Griess Reagent
Griess Reagent
Griess Reagent
Thermo Scientific Chemicals

Griess Reagent

 
Have Questions?
Catalog NumberQuantity
32867050050 mL
Catalog number 328670500
Price (USD)
158.65
Special offer
Online exclusive
Ends: 26-Sep-2025
196.00
Save 37.35 (19%)
Each
In stock
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)
158.65
Special offer
Online exclusive
Ends: 26-Sep-2025
196.00
Save 37.35 (19%)
Each
Add to cart
Specifications
Chemical Name or MaterialGriess Reagent
ColorColorless to Pink
Density1.0300g/mL
Health Hazard 1Danger
Health Hazard 2GHS P Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link