Thermo Scientific Chemicals

NG-Monomethyl-L-arginine monoacetate, 99%

Catalog number: 328720250
25 mg, Glass Bottle, Each
Thermo Scientific Chemicals

NG-Monomethyl-L-arginine monoacetate, 99%

Catalog number: 328720250
25 mg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 328720250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
53308-83-1
IUPAC Name
(2S)-2-amino-5-(N''-methylcarbamimidamido)pentanoic acid; acetic acid
Molecular Formula
C9H20N4O4
InChI Key
IKPNWIGTWUZCKM-JEDNCBNOSA-N
SMILES
CC(O)=O.CN=C(N)NCCC[C@H](N)C(O)=O
Appearance (Form)
Powder
Infrared spectrum
Conforms
TLC
>=98.5 %
Melting point
180°C to 190°C
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text