Thermo Scientific Chemicals

ODQ, 98%, Thermo Scientific Chemicals

Catalog number: 328800500
50 mg, Glass bottle, Each
Thermo Scientific Chemicals

ODQ, 98%, Thermo Scientific Chemicals

Catalog number: 328800500
50 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 328800500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
41443-28-1
IUPAC Name
1H-[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
Molecular Formula
C9H5N3O2
InChI Key
LZMHWZHOZLVYDL-UHFFFAOYSA-N
SMILES
O=C1ON=C2C=NC3=CC=CC=C3N12
Appearance (Color)
Off-white to yellow
Infrared spectrum
Conforms
HPLC
>=97.5 %
Melting point
160°C to 170°C
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text