Thermo Scientific Chemicals

(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride, Thermo Scientific Chemicals

Catalog number: 328871000
100 mg, Each
Thermo Scientific Chemicals

(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride, Thermo Scientific Chemicals

Catalog number: 328871000
100 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 328871000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69056-38-8
IUPAC Name
dihydrogen (6R)-2-amino-6-[(1R,2S)-1,2-dihydroxypropyl]-1,4,5,6,7,8-hexahydropteridin-4-one dichloride
Molecular Formula
C9H17Cl2N5O3
InChI Key
RKSUYBCOVNCALL-NTVURLEBSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].C[C@H](O)[C@H](O)[C@H]1CNC2=C(N1)C(=O)N=C(N)N2
Appearance (Color)
White to light yellow
Appearance (Form)
Powder
Loss on drying
=<2 %
Infrared spectrum
Conforms
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text