(±)-Ibotenic acid monohydrate, 95%, Thermo Scientific Chemicals
(±)-Ibotenic acid monohydrate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(±)-Ibotenic acid monohydrate, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 329130010
Price (USD)/ Each
365.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
430.00 
Save 64.35 (15%)
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
365.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
430.00 
Save 64.35 (15%)
Add to cart
(±)-Ibotenic acid monohydrate, 95%, Thermo Scientific Chemicals
Catalog number329130010
Price (USD)/ Each
365.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
430.00 
Save 64.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-amino-2-(3-oxo-2,3-dihydro-1,2-oxazol-5-yl)acetic acid hydrate
Molecular FormulaC5H8N2O5
InChI KeyQVKQQLYSTODTJN-UHFFFAOYNA-N
SMILESO.NC(C(O)=O)C1=CC(=O)NO1
Molecular Weight (g/mol)176.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
NMRConsistent with structure
HPLC>=94 %
Appearance (Form)Solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only