N-Methyl-D-aspartic acid, 98%, Thermo Scientific Chemicals
N-Methyl-D-aspartic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Methyl-D-aspartic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
Packaging:
Glass bottle
Catalog number 329190500
Price (USD)/ Each
73.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
86.20 
Save 12.55 (15%)
-
Add to cart
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
73.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
86.20 
Save 12.55 (15%)
Add to cart
N-Methyl-D-aspartic acid, 98%, Thermo Scientific Chemicals
Catalog number329190500
Price (USD)/ Each
73.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
86.20 
Save 12.55 (15%)
-
Add to cart
Chemical Identifiers
CAS6384-92-5
IUPAC Name(2R)-2-(methylazaniumyl)butanedioate
Molecular FormulaC5H8NO4
InChI KeyHOKKHZGPKSLGJE-GSVOUGTGSA-M
SMILESC[NH2+][C@H](CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Color)White to off-white
Specific optical rotation-26° to -30° (20°C, 589 nm) (c=1, 6 N HCl)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only