Nifedipine
Thermo Scientific Chemicals

Nifedipine

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 329270050
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
Add to cart
Nifedipine
Catalog number329270050
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Name3,5-dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molecular FormulaC17H18N2O6
InChI KeyHYIMSNHJOBLJNT-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1[N+]([O-])=O)C(=O)OC
Molecular Weight (g/mol)346.34
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=98.0 %
Residue after ignition=<0.1 %
Appearance (Color)Yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only