(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
Thermo Scientific Chemicals

(±)-Verapamil hydrochloride, 99+%

CAS: 152-11-4 | C27H38N2O4·HCl | 491.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 329330010
Price (USD)
98.80
Each
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
98.80
Each
Add to cart
Chemical Identifiers
CAS152-11-4
IUPAC Namehydrogen 2-(3,4-dimethoxyphenyl)-5-{[2-(3,4-dimethoxyphenyl)ethyl](methyl)amino}-2-(propan-2-yl)pentanenitrile chloride
Molecular FormulaC27H39ClN2O4
InChI KeyDOQPXTMNIUCOSY-UHFFFAOYNA-N
SMILES[H+].[Cl-].COC1=CC=C(CCN(C)CCCC(C#N)(C(C)C)C2=CC=C(OC)C(OC)=C2)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point140°C to 144°C
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration with HClO499.0 to 100.5 % (on dried substance)
pH4.5 to 6.5 (5 % solution)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link