Thermo Scientific Chemicals

n-Dodecyl-β-D-maltoside, 99%, high purity, Thermo Scientific Chemicals

Catalog number: 329370050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

n-Dodecyl-β-D-maltoside, 99%, high purity, Thermo Scientific Chemicals

Catalog number: 329370050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 329370050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69227-93-6
IUPAC Name
(2R,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6R)-6-(dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C24H46O11
InChI Key
NLEBIOOXCVAHBD-QKMCSOCLSA-N
SMILES
CCCCCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O
Appearance (Color)
White
Appearance (Form)
Powder
Specific optical rotation
+45° to +49.5° (20°C, 589 nm) (c=1, water)
Infrared spectrum
Conforms
HPLC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text