1-(4-Chlorophenyl)ethylamine, 90%, Thermo Scientific Chemicals
1-(4-Chlorophenyl)ethylamine, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(4-Chlorophenyl)ethylamine, 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 329470050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS6299-02-1
IUPAC Name1-(4-chlorophenyl)ethan-1-amine
Molecular FormulaC8H10ClN
InChI KeyPINPOEWMCLFRRB-UHFFFAOYNA-N
SMILESCC(N)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=88 %
Appearance (Color)Clear yellow
Infrared spectrumConforms
Refractive index1.5455 to 1.5475 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only