Thermo Scientific Chemicals

5-Bromo-2-furaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 329520100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Bromo-2-furaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 329520100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 329520100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1899-24-7
IUPAC Name
5-bromofuran-2-carbaldehyde
Molecular Formula
C5H3BrO2
InChI Key
WJTFHWXMITZNHS-UHFFFAOYSA-N
SMILES
BrC1=CC=C(O1)C=O
Appearance (Color)
White to yellow to light brown
Appearance (Form)
Powder or crystals
GC
>=96.0 %
Melting point
80°C to 85°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text