Brij™ 35, 30 w/v % solution in water, Thermo Scientific Chemicals
Brij™ 35, 30 w/v % solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Brij™ 35, 30 w/v % solution in water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
2.5 L
Catalog number 329580010
Price (USD)/ Each
160.65
Online exclusive
175.00 
Save 14.35 (8%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
160.65
Online exclusive
175.00 
Save 14.35 (8%)
Add to cart
Brij™ 35, 30 w/v % solution in water, Thermo Scientific Chemicals
Catalog number329580010
Price (USD)/ Each
160.65
Online exclusive
175.00 
Save 14.35 (8%)
-
Add to cart
Chemical Identifiers
MDL NumberMFCD00043063
CAS9002-92-0, 7732-18-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Jelly substance
Assay29 to 31 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only