1x1 image pixel for data collection
Brij™ L23-SO-(AP), 30 w/v % sol. in water, previous name Brij™ 35
Brij™ L23-SO-(AP), 30 w/v % sol. in water, previous name Brij™ 35
Brij™ L23-SO-(AP), 30 w/v % sol. in water, previous name Brij™ 35
Brij™ L23-SO-(AP), 30 w/v % sol. in water, previous name Brij™ 35
Brij™ L23-SO-(AP), 30 w/v % sol. in water, previous name Brij™ 35
Thermo Scientific Chemicals

Brij™ L23-SO-(AP), 30 w/v % sol. in water, previous name Brij™ 35

CAS: 9002-92-0 | C58H118O24
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Catalog number 329581000
Price (USD)
60.80
Each
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)
60.80
Each
Add to cart
Chemical Identifiers
CAS9002-92-0
MDL NumberMFCD00043063
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Jelly substance
Assay29 to 31 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link