Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400, Thermo Scientific Chemicals
Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
Catalog number 329690050
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
96.60 
Save 13.95 (14%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
96.60 
Save 13.95 (14%)
Add to cart
Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400, Thermo Scientific Chemicals
Catalog number329690050
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
96.60 
Save 13.95 (14%)
-
Add to cart
Chemical Identifiers
Molecular Formula(C14H11N)n
SMILES*-CC(-*)N1C2=C(C=CC=C2)C2=C1C=CC=C2
MDL NumberMFCD00134336
CAS25067-59-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)Off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only