Cumene, 99.9%, extra pure, Thermo Scientific Chemicals
Cumene, 99.9%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cumene, 99.9%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
5 mL
500 mL
Packaging:
View all
Glass Bottle
Catalog number 329730025
Price (USD)/ Each
76.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.60 
Save 12.95 (14%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
76.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.60 
Save 12.95 (14%)
Add to cart
Cumene, 99.9%, extra pure, Thermo Scientific Chemicals
Catalog number329730025
Price (USD)/ Each
76.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.60 
Save 12.95 (14%)
-
Add to cart
Chemical Identifiers
CAS98-82-8
IUPAC Name(propan-2-yl)benzene
Molecular FormulaC9H12
InChI KeyRWGFKTVRMDUZSP-UHFFFAOYSA-N
SMILESCC(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Density(20°C) 0.861 to 0.863 kg/l
Infrared spectrumConforms
Refractive index1.4900 to 1.4930 (20°C, 589 nm)
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only