Heptadecanoic acid, 98%, Thermo Scientific Chemicals
Heptadecanoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Heptadecanoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 329740250
Price (USD)/ Each
174.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
205.00 
Save 30.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
174.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
205.00 
Save 30.35 (15%)
Add to cart
Heptadecanoic acid, 98%, Thermo Scientific Chemicals
Catalog number329740250
Price (USD)/ Each
174.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
205.00 
Save 30.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameheptadecanoic acid
Molecular FormulaC17H34O2
InChI KeyKEMQGTRYUADPNZ-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCC(O)=O
Molecular Weight (g/mol)270.46
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
GC>=97.5 %
Melting point59°C to 63°C
Appearance (Form)Glistening crystalline powder or flakes
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only