Diphenylsilane, 97%, Thermo Scientific Chemicals
Diphenylsilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenylsilane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 329770050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namediphenylsilane
Molecular FormulaC12H12Si
InChI KeyVDCSGNNYCFPWFK-UHFFFAOYSA-N
SMILES[SiH2](C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)184.31
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5780 to 1.5800 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only