Thermo Scientific Chemicals

Diphenylsilane, 97%, Thermo Scientific Chemicals

Catalog number: 329770250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Diphenylsilane, 97%, Thermo Scientific Chemicals

Catalog number: 329770250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 329770250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
775-12-2
IUPAC Name
diphenylsilane
Molecular Formula
C12H12Si
InChI Key
VDCSGNNYCFPWFK-UHFFFAOYSA-N
SMILES
[SiH2](C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.5780 to 1.5800 (20°C, 589 nm)
Appearance (Color)
Clear colorless
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text