Thermo Scientific Chemicals

Citric acid, for molecular biology, anhydrous, Rnase and Protease free, Thermo Scientific Chemicals

Catalog number: 329860010
1 kg, Each
Thermo Scientific Chemicals

Citric acid, for molecular biology, anhydrous, Rnase and Protease free, Thermo Scientific Chemicals

Catalog number: 329860010
1 kg, Each
Quantity
Catalog number: 329860010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-92-9
IUPAC Name
2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Formula
C6H8O7
InChI Key
KRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILES
OC(=O)CC(O)(CC(O)=O)C(O)=O
Water
=<0.5 % (K.F.)
Dnase,Rnase,Protease activity
none detected
Oxalate (C2O4)
=<500 ppm
Titration with NaOH
>=99 %
Carboniz. sub. by hot H2SO4
(as per ACS) passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for molecular biology
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text