Thermo Scientific Chemicals

N-Hydroxyphthalimide, 98%, Thermo Scientific Chemicals

Catalog number: 329875000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

N-Hydroxyphthalimide, 98%, Thermo Scientific Chemicals

Catalog number: 329875000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 329875000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
524-38-9
IUPAC Name
2-hydroxy-2,3-dihydro-1H-isoindole-1,3-dione
Molecular Formula
C8H5NO3
InChI Key
CFMZSMGAMPBRBE-UHFFFAOYSA-N
SMILES
ON1C(=O)C2=CC=CC=C2C1=O
Appearance (Form)
Powder
Water
=<2 %
Infrared spectrum
Conforms
Appearance (Color)
Yellow
Titration with NaOH
97.5 to 102.5 % (On anhydrous substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text