Thermo Scientific Chemicals

D(+)-Maltose monohydrate, 92+%

Catalog number: 329911000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

D(+)-Maltose monohydrate, 92+%

Catalog number: 329911000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 329911000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6363-53-7
IUPAC Name
2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
Molecular Formula
C12H24O12
InChI Key
HBDJFVFTHLOSDW-UHFFFAOYNA-N
SMILES
O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O
Sulfated ash
=<0.1 %
Specific optical rotation
+135° to +139° (20°C, 589 nm) (c=4, H2O, NH3)
Specific optical rotation
on dry substance
Infrared spectrum
Conforms
HPLC
>=92.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text