Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8 atom % D
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8 atom % D
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8 atom % D
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8 atom % D
Thermo Scientific™

Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8 atom % D

Methanol-d4, CH4O, CAS Number-811-98-3, methan-d3-ol-d, 2h4 methanol, methyl-d3 alcohol-d, perdeuteromethanol, methanol-d4, tetradeuteromethanol, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d, methyl alcohol-d4, cd3od, 50mL, 01, III, 1187, Colorless, 0.8880g/mL, 99.75% min., 212-378-6 | CAS: 811-98-3 | CH4O | 36.066 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
10 mL
25 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 329920250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS811-98-3
IUPAC Name(²H₃)methan(²H)ol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-MZCSYVLQSA-N
SMILES[2H]OC([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Water=<0.05 % (Coulometric)
D-Enrichment>=99.75 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470704Certificate of AnalysisMay 17, 2025329920500, 329920050, 329920250, 329920100
A0471329Certificate of AnalysisMay 17, 2025329920500, 329920050, 329920250, 329920100
A0471719Certificate of AnalysisMay 17, 2025329920500, 329920050, 329920250, 329920100
A0416152Certificate of AnalysisFeb 12, 2025329920500, 329920050, 329920250, 329920100
A0466742Certificate of AnalysisOct 31, 2024329920500, 329920050, 329920250, 329920100
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection