4-chlorobutyronitrile, 97%, Thermo Scientific Chemicals
4-chlorobutyronitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-chlorobutyronitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number 329945000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS628-20-6
IUPAC Name4-chlorobutanenitrile
Molecular FormulaC4H6ClN
InChI KeyZFCFBWSVQWGOJJ-UHFFFAOYSA-N
SMILESClCCCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.4420 to 1.4440 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only