Chemical Identifiers
CAS176763-62-5
IUPAC Name2,4-di-tert-butyl-6-{[(2-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}cyclohexyl)amino]methylidene}cyclohexa-2,4-dien-1-one cobalt
Molecular FormulaC36H54CoN2O2
InChI KeyXJWCTQVLJVAWMC-UHFFFAOYNA-N
SMILES[Co].CC(C)(C)C1=CC(=CNC2CCCCC2NC=C2C=C(C=C(C2=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C
View more