3-(2-Furyl)propanoic acid, 98%
3-(2-Furyl)propanoic acid, 98%
3-(2-Furyl)propanoic acid, 98%
3-(2-Furyl)propanoic acid, 98%
Thermo Scientific Chemicals

3-(2-Furyl)propanoic acid, 98%

CAS: 935-13-7 | C7H8O3 | 140.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
33010010010 gGlass bottle
Catalog number 330100100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS935-13-7
IUPAC Name3-(furan-2-yl)propanoate
Molecular FormulaC7H7O3
InChI KeyXLTJXJJMUFDQEZ-UHFFFAOYSA-M
SMILES[O-]C(=O)CCC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with NaOH>=97.5 %
Appearance (Color)White to brown
Melting point54°C to 58°C
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link