Gibco™

Fibronectin Bovine Protein, Plasma

Catalog number: 33010018
Have Questions?
Gibco™

Fibronectin Bovine Protein, Plasma

Catalog number: 33010018
Have Questions?
Catalog Number
33010018
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
330100181 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Fibronectin is a large, cell surface and plasma protein that exhibits structural and adhesive properties in cell-associated fibrillar matrices. Fibronectin is one of the primary cell adhesion molecules. Bovine Plasma Fibronectin is purified from plasma by affinity chromatography on gelatin-Sepharose and further purified by ion exchange chromatography to remove glycosaminoglycans. Bovine Fibronectin is filtered (0,2 μm) and supplied lyophilized.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Application)
Cell Culture
Quantity
1 mg
Gene Alias
Fibronectin, Bovine Plasma
Species
Cattle/Bovine
Shipping Condition
Approved for shipment at Room Temperature or on Wet or Dry Ice
Form
Solid
Protein Subtype
Fibronectin
Research Category
Extracellular Matrix & Cell Adhesion, Stem Cell Research

Contents & Storage

Store in freezer (-5 to -30°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text