3-(2-Thienyl)propanoic acid, 98%, Thermo Scientific Chemicals
3-(2-Thienyl)propanoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(2-Thienyl)propanoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 330110010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5928-51-8
IUPAC Name3-(thiophen-2-yl)propanoate
Molecular FormulaC7H7O2S
InChI KeyMJPVYTKZYZPIQA-UHFFFAOYSA-M
SMILES[O-]C(=O)CCC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystals or needles
Melting point45°C to 49°C
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only