3-(2-Thienyl)propanoic acid, 98%
3-(2-Thienyl)propanoic acid, 98%
3-(2-Thienyl)propanoic acid, 98%
3-(2-Thienyl)propanoic acid, 98%
Thermo Scientific Chemicals

3-(2-Thienyl)propanoic acid, 98%

CAS: 5928-51-8 | C7H8O2S | 156.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 330110050
Price (USD)
83.65
Online Exclusive
92.30
Save 8.65 (9%)
Each
In stock
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
83.65
Online Exclusive
92.30
Save 8.65 (9%)
Each
Add to cart
Chemical Identifiers
CAS5928-51-8
IUPAC Name3-(thiophen-2-yl)propanoate
Molecular FormulaC7H7O2S
InChI KeyMJPVYTKZYZPIQA-UHFFFAOYSA-M
SMILES[O-]C(=O)CCC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Crystals or needles
Melting point45°C to 49°C
Appearance (Color)White to beige
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0387647Certificate of AnalysisAug 16, 2022330110010, 330110050
A0382845Certificate of AnalysisMar 24, 2022330110010, 330110050
2 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link