Thermo Scientific Chemicals

2-chloromalonaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: 330120500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

2-chloromalonaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: 330120500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 330120500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
36437-19-1
IUPAC Name
(2Z)-2-chloro-3-hydroxyprop-2-enal
Molecular Formula
C3H3ClO2
InChI Key
SYXFXUMRQSTIKC-IWQZZHSRSA-N
SMILES
OC=C(/Cl)C=O
Infrared spectrum
Conforms
Appearance (Color)
Brown
Appearance (Form)
Crystalline powder
Titration with TBAH
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text