Thermo Scientific Chemicals

3,5-Dibromopyridine, 98+%, Thermo Scientific Chemicals

Catalog number: 330130250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3,5-Dibromopyridine, 98+%, Thermo Scientific Chemicals

Catalog number: 330130250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 330130250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
625-92-3
IUPAC Name
3,5-dibromopyridine
Molecular Formula
C5H3Br2N
InChI Key
SOSPMXMEOFGPIM-UHFFFAOYSA-N
SMILES
BrC1=CC(Br)=CN=C1
GC
>=98.0 %
Appearance (Form)
Powder or crystals or needles
Melting point
110°C to 114°C
Appearance (Color)
White to light beige
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text