Benzo[b]furan-2-carboxaldehyde, 96%, Thermo Scientific Chemicals
Benzo[b]furan-2-carboxaldehyde, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzo[b]furan-2-carboxaldehyde, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 330140050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name1-benzofuran-2-carbaldehyde
Molecular FormulaC9H6O2
InChI KeyADDZHRRCUWNSCS-UHFFFAOYSA-N
SMILESO=CC1=CC2=CC=CC=C2O1
Molecular Weight (g/mol)146.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.6300 to 1.6340 (20°C, 589 nm)
GC>=95 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only