1x1 image pixel for data collection
Fibronectin Human Protein, Plasma
Fibronectin Human Protein, Plasma
Gibco™

Fibronectin Human Protein, Plasma

Fibronectin is a large, cell surface and plasma protein that exhibits structural and adhesive properties in cell-associated fibrillar matrices. FibronectinRead more
Have Questions?
Catalog NumberQuantity
330160155 mg
Catalog number 33016015
Price (USD)
634.65
Online Exclusive
690.00
Save 55.35 (8%)
Each
In stock
Add to cart
Quantity:
5 mg
Price (USD)
634.65
Online Exclusive
690.00
Save 55.35 (8%)
Each
Add to cart
Fibronectin is a large, cell surface and plasma protein that exhibits structural and adhesive properties in cell-associated fibrillar matrices. Fibronectin is one of the primary cell adhesion molecules. Human Plasma Fibronectin is isolated from human plasma by affinity chromtography. Fibronectin has a molecular weight of 440,000 and is made up of two 220,000 dalton polypeptides. Human Fibronectin is filtered (0,2UI) and supplied lyophilized.

Performance and Quality Testing
The purity is determined by SDS-PAGE. Bovine and human fibronectins are assayed for their ability to support cell attachment and spreading, and screened to confirm absence of mycoplasma or bacterial contamination.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Cell Culture
Quantity5 mg
Gene AliasFibronectin Human, Plasma
SpeciesHuman
Shipping ConditionRoom Temperature
FormLyophilized
Protein SubtypeFibronectin
Research CategoryExtracellular Matrix & Cell Adhesion, Stem Cell Research
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link