Thermo Scientific Chemicals

Acrylamide / N,N'-Methylenebisacrylamide 19:1, for biochemistry, 40% mix solution in

Catalog number: 330201000
100 mL, Each
Thermo Scientific Chemicals

Acrylamide / N,N'-Methylenebisacrylamide 19:1, for biochemistry, 40% mix solution in

Catalog number: 330201000
100 mL, Each
Quantity
Catalog number: 330201000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
79-06-1, 7732-18-5, 110-26-9, 79-06-1
Appearance (Color)
Clear colorless
Appearance (Form)
Solution
Conductivity
=<100 µS/cm
UV
(5% in water)
UV
at 290 nm A: =<0.7

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text