Acrylamide / N,N'-Methylenebisacrylamide 37.5:1, for biochemistry, 40% mix solution
Acrylamide / N,N'-Methylenebisacrylamide 37.5:1, for biochemistry, 40% mix solution
Thermo Scientific Chemicals

Acrylamide / N,N'-Methylenebisacrylamide 37.5:1, for biochemistry, 40% mix solution

Acrylamide / N,N'-Methylenebisacrylamide, CAS # 79-06-1, 7732-18-5, 110-26-9, 79-06-1, is a water soluble compound that cross-links to produce polyacrylamide gels that are widely used in chromatography and the electrophoretic separation of nucleic acids and proteins.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass Bottle
Catalog number 330225000
Price (USD)/ Each
168.65
Online exclusive
198.00 
Save 29.35 (15%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
168.65
Online exclusive
198.00 
Save 29.35 (15%)
Add to cart
Acrylamide / N,N'-Methylenebisacrylamide 37.5:1, for biochemistry, 40% mix solution
Catalog number330225000
Price (USD)/ Each
168.65
Online exclusive
198.00 
Save 29.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameprop-2-enamide
Molecular FormulaC3H5NO
InChI KeyHRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILESNC(=O)C=C
Molecular Weight (g/mol)71.08
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
UVat 290 nm A: =<0.1
Acrylic acid (HPLC)=<0.001 %
Appearance (Form)Solution
UV(1 % in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Acrylamide / N,N'-Methylenebisacrylamide is a water-soluble, odorless, white crystalline powder
  • This compound polymerizes in the presence of free radicals to generate gels whose pore size may be regulated by manipulating the relative concentrations of this mixture of compounds

Applications

  • Polyacrylamide gels are widely used in biochemistry, forensic chemistry, genetics, molecular biology, and biotechnology to separate proteins or nucleic acids, according to their electrophoretic mobility
  • Because of their higher resolution compared to other gels, polyacrylamide gels are also used in the analysis of smaller RNA species from mixed samples
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only