Thermo Scientific Chemicals

1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 330360250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 330360250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 330360250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2050-77-3
IUPAC Name
1-iododecane
Molecular Formula
C10H21I
InChI Key
SKIDNYUZJPMKFC-UHFFFAOYSA-N
SMILES
CCCCCCCCCCI
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Stabilizer
copper
Refractive index
1.4840 to 1.4870 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text