Thermo Scientific Chemicals

Antimony pentasulfide, pure, Thermo Scientific Chemicals

Catalog number: 330410010
1 kg, Each
Thermo Scientific Chemicals

Antimony pentasulfide, pure, Thermo Scientific Chemicals

Catalog number: 330410010
1 kg, Each
Quantity
Catalog number: 330410010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1315-04-4
IUPAC Name
[(disulfanylidenestibanyl)sulfanyl]stibanedithione
Molecular Formula
S5Sb2
InChI Key
PPKVREKQVQREQD-UHFFFAOYSA-N
SMILES
S=[Sb](=S)S[Sb](=S)=S
Assay
>=54 % (Sb)
Appearance (Color)
Yellow to orange-red
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text